
6-Bromo-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylic acid
Catalog # APOSPC300742-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:309.039 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1427460-57-8
Formula:C₉H₄BrF₃N₂O₂
MDL Number:MFCD18804153