Order Entry
United Kingdom
ContactUsLinkComponent
Ethyl 1-{[2-methyl-6-(trifluoromethyl)pyridin-3-yl]methyl}piperidine-3-carboxylate
Ethyl 1-{[2-methyl-6-(trifluoromethyl)pyridin-3-yl]methyl}piperidine-3-carboxylate
Catalog # APOSPC300603-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 1-{[2-methyl-6-(trifluoromethyl)pyridin-3-yl]methyl}piperidine-3-carboxylate
Catalog # APOSPC300603-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :330.345 g/mol
Storage Temperature:Ambient
CAS Number:1308645-62-6
Formula:C₁₆H₂₁F₃N₂O₂
MDL Number:MFCD13194592