Order Entry
United Kingdom
ContactUsLinkComponent
6-(Trifluoromethyl)-2H-1,4-benzoxazin-3(4H)-one 95%
6-(Trifluoromethyl)-2H-1,4-benzoxazin-3(4H)-one 95%
Catalog # APOSPC300507-1G
Supplier:  Apollo Scientific
CAS Number:  
6-(Trifluoromethyl)-2H-1,4-benzoxazin-3(4H)-one 95%
Catalog # APOSPC300507-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :217.145 g/mol
Melting Pt:152…156 °C
Storage Temperature:Ambient
CAS Number:189940-04-3
Formula:C₉H₆F₃NO₂
MDL Number:MFCD02648641