Order Entry
United Kingdom
ContactUsLinkComponent
{2-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]ethyl}(methoxy)amine, tech
{2-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]ethyl}(methoxy)amine, tech
Catalog # APOSPC28021-1G
Supplier:  Apollo Scientific
CAS Number:  
{2-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]ethyl}(methoxy)amine, tech
Catalog # APOSPC28021-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :254.637 g/mol
Storage Temperature:Ambient
CAS Number:400078-95-7
Formula:C₉H₁₀ClF₃N₂O
MDL Number:MFCD02571791