Order Entry
United Kingdom
ContactUsLinkComponent
 
1-[2-(4-Fluorophenyl)cyclopropyl]ethan-1-one 95%
Catalog # APOSPC28004-1G
Supplier:  Apollo Scientific
CAS Number:  
1-[2-(4-Fluorophenyl)cyclopropyl]ethan-1-one 95%
Catalog # APOSPC28004-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :178.203 g/mol
Storage Temperature:Ambient
CAS Number:338392-56-6
Formula:C₁₁H₁₁FO
MDL Number:MFCD01315497