
4-Fluoro-2-(trifluoromethyl)phenylhydrazine hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:502496-21-1
:
Formula:C₇H₆F₄N₂·ClH
MDL Number:MFCD03094163
:230.59 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together