Order Entry
United Kingdom
ContactUsLinkComponent
2-Amino-5,6-bis(difluoromethoxy)-1,3-benzothiazole 98%
Irritant
2-Amino-5,6-bis(difluoromethoxy)-1,3-benzothiazole 98%
Catalog # APOSPC2205-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-Amino-5,6-bis(difluoromethoxy)-1,3-benzothiazole 98%
Catalog # APOSPC2205-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :282.215 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:887268-01-1
Formula:C₉H₆F₄N₂O₂S
MDL Number:MFCD04972671