Methyl 4-amino-3-fluoro-5-nitro-2-(phenylamino)benzoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:606093-58-7
:
Formula:C₁₄H₁₂FN₃O₄
MDL Number:MFCD09909911
:305.261 g/mol
Storage Temperature:Ambient
Frequently Bought Together