
2-Amino-3'-fluorobiphenyl hydrochloride
Catalog # APOSPC11124-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:223.674 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:139769-18-9
Formula:C₁₂H₁₀FN·ClH
MDL Number:MFCD06795781