
3-Bromo-4-fluorobenzoyl chloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:672-75-3
:
Formula:C₇H₃BrClFO
MDL Number:MFCD04972690
:237.454 g/mol
Boiling Pt:100…112 °C (11 mm Hg)
Melting Pt:43…45 °C
Flash Pt:>110 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
Frequently Bought Together