Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Fluoro-2-thiobenzoic acid
5-Fluoro-2-thiobenzoic acid
5-Fluoro-2-thiobenzoic acid
APOSPC0950-1G
:
Apollo Scientific
:
5-Fluoro-2-thiobenzoic acid
APOSPC0950-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
172.177 g/mol
:
137…142 °C
:
>110 °C
:
GHS07
:
Refrigerator
:
120121-07-5
:
C₇H₅FO₂S
:
MFCD02683508