
2,4-Dibromo-6-fluorobenzaldehyde
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:205683-34-7
:
Formula:C₇H₃Br₂FO
MDL Number:MFCD03094089
:281.905 g/mol
Melting Pt:57…60 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together