Order Entry
United Kingdom
ContactUsLinkComponent
2-Cyclopropyl-3-nitro-1H-pyrazole 98%
2-Cyclopropyl-3-nitro-1H-pyrazole 98%
Catalog # APOSOR962920-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Cyclopropyl-3-nitro-1H-pyrazole 98%
Catalog # APOSOR962920-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :153.139 g/mol
Storage Temperature:Ambient
CAS Number:1249581-67-6
Formula:C₆H₇N₃O₂
MDL Number:MFCD16249530