3H-Imidazo[4,5-c]pyridine-6-carboxylic acid methyl ester 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:82523-07-7
:
Formula:C₈H₇N₃O₂
MDL Number:MFCD11656494
:177.16 g/mol
Storage Temperature:Ambient
Frequently Bought Together