Order Entry
United Kingdom
ContactUsLinkComponent
(4-Cyanophenoxy)acetic acid 96%
(4-Cyanophenoxy)acetic acid 96%
Catalog # APOSOR962557-5G
Supplier:  Apollo Scientific
CAS Number:  
(4-Cyanophenoxy)acetic acid 96%
Catalog # APOSOR962557-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :177.157 g/mol
Storage Temperature:Ambient
CAS Number:1878-82-6
Formula:C₉H₇NO₃
MDL Number:MFCD01250758