Order Entry
United Kingdom
ContactUsLinkComponent
4-(Isopropylsulphanyl)benzaldehyde 98%
4-(Isopropylsulphanyl)benzaldehyde 98%
Catalog # APOSOR960283-1G
Supplier:  Apollo Scientific
CAS Number:  
4-(Isopropylsulphanyl)benzaldehyde 98%
Catalog # APOSOR960283-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :180.267 g/mol
Storage Temperature:Ambient
CAS Number:84264-99-3
Formula:C₁₀H₁₂OS
MDL Number:MFCD18887933