Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(2-Methylpropyl)pyrazole-3-carbaldehyde
2-(2-Methylpropyl)pyrazole-3-carbaldehyde 95%
2-(2-Methylpropyl)pyrazole-3-carbaldehyde 95%
2-(2-Methylpropyl)pyrazole-3-carbaldehyde 95%
APOSOR959892-1G
:
Apollo Scientific
:
2-(2-Methylpropyl)pyrazole-3-carbaldehyde 95%
APOSOR959892-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
152.194 g/mol
:
Ambient
:
1236366-12-3
:
C₈H₁₂N₂O
:
MFCD21609707