Order Entry
United Kingdom
ContactUsLinkComponent
3-(3-Hydroxy-phenyl)-acrylic acid ethyl ester 95%
3-(3-Hydroxy-phenyl)-acrylic acid ethyl ester 95%
Catalog # APOSOR957724-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(3-Hydroxy-phenyl)-acrylic acid ethyl ester 95%
Catalog # APOSOR957724-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :192.211 g/mol
Storage Temperature:Ambient
CAS Number:96251-92-2
Formula:C₁₁H₁₂O₃
MDL Number:MFCD00184677