Order Entry
United Kingdom
ContactUsLinkComponent
1-(2-Chlorobenzyl)pyrrolidine 95%
1-(2-Chlorobenzyl)pyrrolidine 95%
Catalog # APOSOR957031-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(2-Chlorobenzyl)pyrrolidine 95%
Catalog # APOSOR957031-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :195.689 g/mol
Storage Temperature:Ambient
Formula:C₁₁H₁₄ClN
MDL Number:MFCD02661502