Order Entry
United Kingdom
ContactUsLinkComponent
Methyl quinoline-4-carboxylate 97%
Methyl quinoline-4-carboxylate 97%
Catalog # APOSOR955585-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl quinoline-4-carboxylate 97%
Catalog # APOSOR955585-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :187.195 g/mol
Storage Temperature:Ambient
CAS Number:21233-61-4
Formula:C₁₁H₉NO₂
MDL Number:MFCD01366551