(3,3-Dimethylcyclobutyl)methanamine
Bottle, 1 g
Catalog # APOSOR948833-1G
Supplier: Apollo Scientific
(3,3-Dimethylcyclobutyl)methanamine
Catalog # APOSOR948833-1G
Supplier: Apollo Scientific
CAS Number: 1244949-67-4
Jump to:
Product Details & Documents
- CAS Number:1244949-67-4
- Formula:C₇H₁₅N
- MDL Number:MFCD17170418
- Molecular Weight:113.201 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:1 g



