3-[(4-Oxo-3,4-dihydro-1-phthalazinyl)methyl]benzoic acid 95%
Catalog # APOSOR946199-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:280.278 g/mol
Storage Temperature:Ambient
CAS Number:420846-72-6
Formula:C₁₆H₁₂N₂O₃
MDL Number:MFCD20483902