2-Cyclohexyl-5-methyl-2H-pyrazol-3-ol 95%
Supplier: Apollo Scientific
Certificates
About this item
:
Formula:C₁₀H₁₆N₂O
MDL Number:MFCD01332424
:180.247 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together