Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Methyl-2-nitrobenzaldehyde
4-Methyl-2-nitrobenzaldehyde 95%
4-Methyl-2-nitrobenzaldehyde 95%
APOSOR930693-1G
:
Apollo Scientific
:
4-Methyl-2-nitrobenzaldehyde 95%
APOSOR930693-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
165.146 g/mol
:
Ambient
:
20357-22-6
:
C₈H₇NO₃
:
MFCD08669895