3,4-Dihydro-2H-quinoline-1-carboxylic acid methyl ester 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:94567-78-9
:
Formula:C₁₁H₁₃NO₂
MDL Number:MFCD03364950
:191.226 g/mol
Storage Temperature:Ambient
Frequently Bought Together