3-Bromo-4-formylbenzonitrile 97%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 2-Bromo-4-cyanobenzaldehyde
CAS Number:89891-69-0
:
Formula:C₈H₄BrNO
MDL Number:MFCD09261031
:210.027 g/mol
Melting Pt:121…124 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together