Order Entry
United Kingdom
ContactUsLinkComponent
6-Methoxy-1-naphthol 95%
6-Methoxy-1-naphthol 95%
Catalog # APOSOR916643-5G
Supplier:  Apollo Scientific
CAS Number:  
6-Methoxy-1-naphthol 95%
Catalog # APOSOR916643-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :174.196 g/mol
Storage Temperature:Ambient
CAS Number:22604-07-5
Formula:C₁₁H₁₀O₂
MDL Number:MFCD03695468