Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Methoxy-1H-indole-3-carboxylic acid
6-Methoxy-1H-indole-3-carboxylic acid 97%
6-Methoxy-1H-indole-3-carboxylic acid 97%
APOSOR913080-5G
:
Apollo Scientific
:
6-Methoxy-1H-indole-3-carboxylic acid 97%
APOSOR913080-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
191.183 g/mol
:
Ambient
:
90924-43-9
:
C₁₀H₉NO₃
:
MFCD05664010