2-Amino-4-bromo-5-chlorophenol ≥97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1037298-14-8
:
Formula:C₆H₅BrClNO
MDL Number:MFCD25978069
Molecular Weight:222.467 g/mol
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together