Order Entry
United Kingdom
ContactUsLinkComponent
tert-Butyl 4-bromo-1-naphthoate 96%
Irritant
tert-Butyl 4-bromo-1-naphthoate 96%
Catalog # APOSOR8570-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
tert-Butyl 4-bromo-1-naphthoate 96%
Catalog # APOSOR8570-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :307.182 g/mol
Melting Pt:25…28 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:929000-22-6
Formula:C₁₅H₁₅BrO₂
MDL Number:MFCD09258636