2-Bromo-5-methoxy-N,N-dimethylbenzylamine
Catalog # APOSOR7801-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:244.128 g/mol
Storage Temperature:Ambient
CAS Number:10126-37-1
Formula:C₁₀H₁₄BrNO
MDL Number:MFCD06657799