


4-(Thien-2-yl)aniline hydrochloride
Catalog # APOSOR7585-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:211.711 g/mol
Melting Pt:185…187 °C
Storage Temperature:Refrigerator
UN:2811
ADR:6.1,III
:GHS05
:GHS06
:GHS07
CAS Number:1172363-16-4
Formula:C₁₀H₉NS·ClH
MDL Number:MFCD00136945