
4-Amino-2'-methylbiphenyl hydrochloride
Catalog # APOSOR7496-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:219.71 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:811842-48-5
Formula:C₁₃H₁₃N·ClH
MDL Number:MFCD06739442