Order Entry
United Kingdom
ContactUsLinkComponent
2'-(Benzyloxy)-[1,1'-biphenyl]-2-amine hydrochloride
Irritant
2'-(Benzyloxy)-[1,1'-biphenyl]-2-amine hydrochloride
Catalog # APOSOR7321-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2'-(Benzyloxy)-[1,1'-biphenyl]-2-amine hydrochloride
Catalog # APOSOR7321-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :311.805 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1170524-29-4
Formula:C₁₉H₁₇NO·ClH
MDL Number:MFCD07366472