
Ethyl 2-sulphanyl-1H-imidazole-4-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:64038-64-8
:MFCD02091509
Formula:C₆H₈N₂O₂S
MDL Number:MFCD00068042
:172.205 g/mol
Melting Pt:191…193 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together