
6-Chloro-2-methyl-3H-imidazo[4,5-c]pyridine
Catalog # APOSOR61532-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:167.596 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:7205-42-7
Formula:C₇H₆ClN₃
MDL Number:MFCD18256899