
2-(Aminomethyl)-3-bromo-5-chloropyridine hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1257535-43-5
:
Formula:C₆H₆BrClN₂·ClH
MDL Number:MFCD17214190
:257.943 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together