(1S,2S)-2-(Ethoxycarbonyl)cyclopropane-1-carboxylic acid 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:175415-95-9
:
Formula:C₇H₁₀O₄
MDL Number:MFCD18089853
:158.152 g/mol
Melting Pt:58…60 °C
Storage Temperature:Refrigerator
Frequently Bought Together