2-Hydrazino-5-methylpyridine 95%
Catalog # APOSOR60226-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:123.156 g/mol
Boiling Pt:90…95 °C (0.03 mm Hg)
Melting Pt:67…72 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:4931-01-5
Formula:C₆H₉N₃
MDL Number:MFCD03425577