
6-Bromo-8-methoxy-2-oxo-2H-chromene-3-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:119686-34-9
:
Formula:C₁₁H₇BrO₅
MDL Number:MFCD01764559
:299.074 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together