
Benzyl 6-oxa-3-azabicyclo[3.1.0]hexane-3-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:31865-25-5
:
Formula:C₁₂H₁₃NO₃
MDL Number:MFCD09608058
:219.237 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together