Order Entry
United Kingdom
ContactUsLinkComponent
3,5-Diamino-6-chloropyrazine-2-carboxylic acid
Irritant
3,5-Diamino-6-chloropyrazine-2-carboxylic acid
Catalog # APOSOR51902-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3,5-Diamino-6-chloropyrazine-2-carboxylic acid
Catalog # APOSOR51902-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :188.572 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:4878-36-8
Formula:C₅H₅ClN₄O₂
MDL Number:MFCD06208349