Order Entry
United Kingdom
ContactUsLinkComponent
5-Amino-3-chloropyrazine-2-carbonitrile ≥97%
5-Amino-3-chloropyrazine-2-carbonitrile ≥97%
Catalog # APOSOR510087-250MG
Supplier:  Apollo Scientific
CAS Number:  
5-Amino-3-chloropyrazine-2-carbonitrile ≥97%
Catalog # APOSOR510087-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :154.557 g/mol
Storage Temperature:Refrigerator
CAS Number:34617-65-7
Formula:C₅H₃ClN₄
MDL Number:MFCD10697777