Order Entry
United Kingdom
ContactUsLinkComponent
2-(Isopropylthio)aniline hydrochloride ≥98%
2-(Isopropylthio)aniline hydrochloride ≥98%
Catalog # APOSOR510028-250MG
Supplier:  Apollo Scientific
CAS Number:  
2-(Isopropylthio)aniline hydrochloride ≥98%
Catalog # APOSOR510028-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :203.732 g/mol
Storage Temperature:Refrigerator
CAS Number:861343-73-9
Formula:C₉H₁₃NS·ClH
MDL Number:MFCD11841393