Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Methyl-1H-indole-5-carbonitrile
3-Methyl-1H-indole-5-carbonitrile ≥97%
3-Methyl-1H-indole-5-carbonitrile ≥97%
APOSOR510006-250MG
:
Apollo Scientific
:
3-Methyl-1H-indole-5-carbonitrile ≥97%
APOSOR510006-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
156.184 g/mol
:
Refrigerator
:
3613-06-7
:
C₁₀H₈N₂
:
MFCD15523476