
(6-Chloropyrazin-2-yl)methanamine hydrochloride 95+%
Catalog # APOSOR50482-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:180.035 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1357945-24-4
Formula:C₅H₆ClN₃·ClH
MDL Number:MFCD20327986