Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromo-1-(2-chloropyridin-4-yl)ethanone
2-Bromo-1-(2-chloropyridin-4-yl)ethanone 90%
2-Bromo-1-(2-chloropyridin-4-yl)ethanone 90%
2-Bromo-1-(2-chloropyridin-4-yl)ethanone 90%
APOSOR480854-1G
:
Apollo Scientific
:
2-Bromo-1-(2-chloropyridin-4-yl)ethanone 90%
APOSOR480854-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
234.478 g/mol
:
55…56.5 °C
:
Ambient
:
2923
:
8+6.1,III
:
GHS05
:
GHS07
:
23794-16-3
:
C₇H₅BrClNO
:
MFCD09702373