Ethyl 4-[1-(prop-2-ynylamino)ethyl]benzoate hydrochloride
Catalog # APOSOR475111-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:267.751 g/mol
Storage Temperature:Ambient
Formula:C₁₄H₁₇NO₂·ClH
MDL Number:MFCD31695652