N-[(1S)-1-(2-Bromophenyl)ethyl]aniline hydrochloride
Catalog # APOSOR475011-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:312.633 g/mol
Storage Temperature:Ambient
Formula:C₁₄H₁₄BrN·ClH
MDL Number:MFCD31695650