Methyl 3-[Hydroxy(phenyl)methyl]isoxazole-4-carboxylate
Catalog # APOSOR471411-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:233.22 g/mol
Storage Temperature:Ambient
CAS Number:2006277-12-7
Formula:C₁₂H₁₁NO₄
MDL Number:MFCD28977379